missing translation for 'onlineSavingsMsg'
Learn More
Ethyl chloroacetate, 99%, Thermo Scientific Chemicals Product Code.: 11462997

Ethyl chloroacetate, 99%, Thermo Scientific Chemicals

Product Code. 11462997
100 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11462997

Brand: Thermo Scientific Alfa Aesar A15554.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ethyl chloroacetate is a reagent used in the preparation of 5 member heterocycles. It is used as pharmaceutical and organic intermediate. It is used as a solvent for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl chloroacetate is a reagent used in the preparation of 5 member heterocycles. It is used as pharmaceutical and organic intermediate. It is used as a solvent for organic synthesis.

Solubility
Soluble in water 20 g/L (20°C).

Notes
Incompatible with acids, bases, oxidizing agents, reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-39-5
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.548
MDL Number MFCD00000932
InChI Key VEUUMBGHMNQHGO-UHFFFAOYSA-N
Synonym ethyl chloroacetate, ethyl chloracetate, ethyl monochloroacetate, acetic acid, chloro-, ethyl ester, chloroacetic acid ethyl ester, ethyl monochloracetate, ethyl chloroethanoate, ethyl alpha-chloroacetate, 2-chloroacetic acid ethyl ester, unii-6h07teh4tfShow MoreShow Less
PubChem CID 7751
IUPAC Name ethyl 2-chloroacetate
SMILES CCOC(=O)CCl

Specifications

Melting Point -26°C
Density 1.159
Boiling Point 142°C to 144°C
Flash Point 54°C (149°F)
Odor Pungent
Linear Formula ClCH2CO2CH2CH3
Refractive Index 1.421
Quantity 100 g
UN Number UN1181
Beilstein 506455
Merck Index 14,3783
Solubility Information Soluble in water 20g/L (20°C).
Formula Weight 122.55
Percent Purity 99%
Chemical Name or Material Ethyl chloroacetate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10245032 Certificate TypeCertificate of Analysis Date16/12/2022 Catalog NumberA15554.22,A15554.36,A15554.0E
Lot Number10236219 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15554.22,A15554.36,A15554.0E
Lot Number10203519 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15554.22,A15554.36,A15554.0E
Lot Number10219452 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15554.22,A15554.36,A15554.0E
Lot Number10106226 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15554.22,A15554.36,A15554.0E
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethyl chloroacetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Ethyl chloroacetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl chloroacetate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.