missing translation for 'onlineSavingsMsg'
Learn More

Eutech™ pH Buffers and Solutions

Eutech™ offers a wide range of pH buffers and solutions for use with pH meters and electrodes

Brand:  Thermo Scientific Eutech 01X211211

13.02 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.



 View more versions of this product

Product Code. 10759503

  • 17.39 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The measure of a liquid's acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications.

Eutech™ offers a wide range of pH buffers and solutions for use with pH meters and electrodes.

Features:

  • Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
  • TRUSTED_SUSTAINABILITY
Specifications

Specifications

Buffer Solution
1.68
Buffer
pH Meter and Electrode
480 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

Supplemental information
  • MIXTURE LIST-Contains : potassium tetraoxalate dihydrate

SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Eutech™ pH Buffers and Solutions >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.