Learn More
Eutech™ pH Buffers and Solutions
Eutech™ offers a wide range of pH buffers and solutions for use with pH meters and electrodes
Brand:  Thermo Scientific 01X638901
 View more versions of this product
Product Code. 15611717
Description
Description
The measure of a liquid's acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications.
Eutech™ offers a wide range of pH buffers and solutions for use with pH meters and electrodes.
Features:
  • Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
  • TRUSTED_SUSTAINABILITY
Specifications
Specifications
Undesignated
pH Meter and Electrode
Liquid
pH Buffer
pH buffer kit, clear buffer solutions each.
4, 7, 10
480 mL
Kit with 480mL bottles
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Eutech™ pH Buffers and Solutions >