Learn More
CleanSpace™ EX Power System
Intrinsically safe battery-powered, positive pressure air-purifying respirator (PAPR). Suitable for use in certain ignitable or potentially explosive atmospheres where there is a risk of explosion.
Brand: CleanSpace™ PAF-0060
This item is not returnable.
View return policy
Description
- Intrinsically safe
- Light weight and comfortable
- High visibility yellow casing for easy recognition
- AirSensit System™ for mask pressure control and breath responsive airflow
- One-button system and standby and auto-start/stop mode for quick don/doffing
- Easy to use support harness
- Altitude compensated filter blocked detection
- Long operating time: up to 8 hours and quick battery recharging: < 2hours
- Easy and accurate flow capability check
Specifications
PAPR Assembly, No Headtop | |
PAF-0060 |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
CleanSpace™ EX Power System > Power Unit
Mask sold separately
Spot an opportunity for improvement?Share a Content Correction