Learn More
Lenz Laborglasintrumente Expansion Adapters with Ground Joints
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01502414
Product Code. 12674873
  • 20.00 € / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12257
  • Conical ground sockets and cones
  • Made of DURAN™ tubing
Specifications
Specifications
Duran Glass
Socket NS 24/29, Cone NS 14/23
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasintrumente Expansion Adapters with Ground Joints > Socket NS 24/29, Cone NS 14/23