missing translation for 'onlineSavingsMsg'
Learn More

Koettermann™ Exploris™ Tall Storage Cabinet

Features soundproof doors.

Brand:  Koettermann™ 507-00016

Product Code. 14885401

  • 2320.00 € / Each
Estimated Shipment: 23-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Used where high load capacity is needed
  • Melamine Topresist™ worktop
Specifications

Specifications

75 cm
Stand alone bench
One steel shelf
120 cm
Trespa TopLab Melamine
75 cm
120 cm
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Koettermann™ Exploris™ Tall Storage Cabinet > Trespa Toplab worktop; D x W x H: 75 x 120 x 75cm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.