Learn More
Gibco™ CD Hybridoma AGT™ Medium
Animal origin-free, chemically defined, protein-free medium
Brand: Gibco™ 12372017
1188.00 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Gibco CD Hybridoma Medium is specifically developed for the ability to support the growth of hybridomas for antibody production.
- Ability to support suspension and stationary hybridomas and myelomas
- Formulated without L-glutamine for stability
- Animal origin-free, protein-free and chemically defined formulation
- Convenient, easy-to-use Advanced Granulation Technology (AGT™) format
Ability to support suspension and stationary hybridomas and myelomas
- Suitable for the culture of recombinant myeloma lines as well as traditional hybridomas, Gibco CD Hybridoma Medium provides high cell densities and IgG production
- Cholesterol-dependent cultures such as NS0 require the addition of 250X Cholesterol Lipid Concentrate
- Gibco CD Hybridoma Medium requires the addition of 8 mM L-glutamine or GlutaMAX™ supplement before use
- GlutaMAX supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format
- Gibco CD Hybridoma Medium is animal origin-free, protein-free and chemically defined, allowing for easier purification of your protein of interest
- Gibco chemically defined media contain no proteins, hydrolysates or components of unknown composition
- The AGT format is an easy-to-use dry powder format. The granules dissolve instantly and pH-adjusts for fast media preparation
- Complete reconstitution instructions are available in the product manual
Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Wet ice

Specifications
Myeloma Cells, Hybridomas | |
Powder (AGT) | |
Serum-free | |
Sodium Pyruvate, Sodium Bicarbonate | |
AGT™, Gibco™ | |
Wet Ice | |
Storage conditions: 2-8°C, dark and dry Shipping conditions: Gel packs Shelf life: 24 months from date of manufacture |
Animal Origin-free, Chemically-defined, Protein-free | |
Hybridoma Medium | |
Non-sterile | |
No Glutamine, No Phenol Red | |
10 L | |
≤10 EU/mL, ≤ 10 EU/ml |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3083049 | Certificate of Analysis | 30/01/2025 | 12372003,12372025,12372001,12372017 |
2387907 | Certificate of Analysis | 29/01/2025 | 12372003,12372025,12372001,12372017 |
2481054 | Certificate of Analysis | 29/01/2025 | 12372003,12372025,12372001,12372017 |
2519529 | Certificate of Analysis | 29/01/2025 | 12372003,12372025,12372001,12372017 |
2531012 | Certificate of Analysis | 29/01/2025 | 12372003,12372025,12372001,12372017 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.