Learn More
Fisherbrand™ Connecting Cable
Connecting cable with BNC Plug for combined ion-sensitive electrode with solid membranes that is ideal for measuring agriculture, water analysis, and calibration of ionic electrodes.
Brand:  Fisherbrand™ 9001041-010B
 View more versions of this product
Product Code. 11776398
  • 44.70 € / Each
Estimated Shipment: 23-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Connecting Cable with BNC Plug
  • For combined ion-sensitive electrode with solid membranes
  • For use with measuring agriculture, water analysis, and calibration of ionic electrodes
Specifications
Specifications
BNC
Connecting Cable
For connecting the electrode to a pH meter, using an anti -static coaxial cable with the correct plug .
3 m
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Connecting Cable > Connector Type: BNC