Learn More
Memmert™ Extended Overtemperature Protection for UF30
Extended overtemperature protection with integrated Pt100 sensor for independent temperature monitoring.
Brand: Memmert™ UF30A6
This item is not returnable.
View return policy
Description
- Accessory
- With integrated Pt100 sensor
- For models with SingleDISPLAY
Specifications
UF30 Oven | |
Integrated Pt100 Sensor |
Product Content Correction
Product Title
Memmert™ Extended Overtemperature Protection for UF30 > For Use With: UF30
Spot an opportunity for improvement?Share a Content Correction