Learn More
Memmert™ Extension of DAkkS Factory Calibration for UF30
Extension of DAkkS factory calibration by an additional freely selectable temperature for Memmert Ovens.
Brand:  Memmert™ UF30E39697
Product Code. 17539245
  • 979.00 € / Each
Explore more special offers
Description
Description
  • Extension of DAkkS calibration at Memmert factory by an additional freely selectable temperature value according to method C (DKD-R 5-7)
Specifications
Specifications
UF30 Oven
Factory Calibration Extension
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ Extension of DAkkS Factory Calibration for UF30 > For Use With: UF30