Learn More
Lenz Laborglasintrumente Extractors Acc. to Soxhlet with Dimroth Condenser
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05360043
This item is not returnable.
View return policy
Description
- The method allows an effective separation with relatively small amounts of extracting solvents.
- Extractor in compact and robust design, with flask connection NS 29/32
- Made of DURAN™ tubing
- Dimroth condenser with thread GL 14
- Round bottom flask
Specifications
150 mL | |
Duran Glass | |
NS 45/40 |
Product Content Correction
Product Title
Lenz Laborglasintrumente Extractors Acc. to Soxhlet with Dimroth Condenser > 150 mL
Spot an opportunity for improvement?Share a Content Correction