Learn More
Falcon™ Round-Bottom Polystyrene Test Tubes
Constructed of optically clear polystyrene for easy viewing of sample. Falcon™ Round-Bottom Polystyrene Tubes are widely used and referenced in laboratory protocols.
104.12 € - 372.24 €
Specifications
Color | Clear |
---|---|
Description | 1400 x G |
Material | Polystyrene |
Disposable | Single-use |
Graduated | No |
Product Code | Brand | Capacity (Metric) | Closure Type | Sterility | Packaging | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Type | Sterility | Packaging | Price | Quantity & Availability | ||||||
10694331
![]() ![]() |
Falcon™
352045 |
19 mL | Screw Cap | Sterile | Individually wrapped, 500/Cs. |
221.29 €
Pack of 500 |
In Stock
Log in to see stock. |
||||||
12678475
![]() ![]() |
Falcon™
352037 |
16 mL | Screw Cap | Sterile | Individually wrapped, 500/Cs. |
217.68 €
Pack of 500 |
Estimated Shipment: 09-09-2025
Log in to see stock. |
||||||
10769861
![]() ![]() |
Falcon™
352025 |
16 mL | Screw Cap | Sterile | Bag of 125, 1000/Cs. |
371.29 €
Pack of 1000 |
In Stock
Log in to see stock. |
||||||
10186360
![]() ![]() |
Falcon™
352008 |
5 mL | Not Included | Non-sterile | Bulk, 1000/Cs. |
104.12 €
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
||||||
10100151
![]() |
Falcon™
352058 |
5 mL | Snap Cap | Sterile | Bag of 25, 500/Cs. |
123.67 €
Pack of 500 |
In Stock
Log in to see stock. ![]() |
||||||
10252421
![]() |
Falcon™
352057 |
14 mL | Snap Cap | Sterile | Bag of 25, 500/Cs. |
135.35 €
Pack of 500 |
In Stock
Log in to see stock. |
||||||
10568531
|
Falcon™
352057 |
14 mL | Snap Cap | Sterile | Bag of 25, 500/Cs. | N/A | N/A | N/A | |||||
10186400
![]() |
Falcon™
352054 |
5 mL | Snap Cap | Sterile | Bag of 125, 1000/Cs. |
222.28 €
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
||||||
10222901
![]() ![]() |
Falcon™
352027 |
8 mL | Screw Cap | Sterile | Bag of 125, 1000/Cs. |
372.24 €
Pack of 1000 |
In Stock
Log in to see stock. |
||||||
10481751
![]() ![]() |
Falcon™
352051 |
14 mL | Snap Cap | Sterile | Bag of 125, 1000/Cs. |
227.79 €
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
||||||
10579511
![]() |
Falcon™
352052 |
5 mL | Not Included | Sterile | 1000/Cs. |
155.70 €
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
||||||
10088710
![]() ![]() |
Falcon™
352003 |
5 mL | Snap Cap | Sterile | Individually wrapped, 500/Cs. |
143.23 €
Pack of 500 |
In Stock
Log in to see stock. |
||||||
10408851
![]() ![]() |
Falcon™
352017 |
14 mL | Not Included | Sterile | Bag of 125, 1000/Cs. |
162.66 €
Pack of 1000 |
In Stock
Log in to see stock. |
||||||
10769331
![]() ![]() |
Falcon™
352001 |
14 mL | Snap Cap | Sterile | Individually wrapped, 500/Cs. |
156.23 €
Pack of 500 |
In Stock
Log in to see stock. |
||||||
Description
- Widely used and referenced in laboratory protocols
- Dual position snap-cap prevents sample loss with ease of handling; provides a secure, positive seal (except for items 352008, 352017, and 352052 which do not have caps)
- Sterile, nonpyrogenicity tested to less than 0.1 EU/mL (Cat. No. 352008 nonsterile)
The foundation for consistent research results
- Provide reliable containment of laboratory fluid samples
- Polystyrene tubes are best suited for procedures requiring high optical clarity
- Heavier gauge walls; unique construction
- Medical-style packaging materials
- 1,400 RCF rating (RCF claims refer to Relative Centrifugal Force measured in g-force for materials with a specific gravity of 1.0, used in an appropriate rotor with correct cushion and safety precautions. Tubes used with organic solvents at temperatures below 0°C may have lower RCF ratings.)
- Tube dimensions and volumes are approximate
Specifications
Clear | |
Polystyrene | |
No |
1400 x G | |
Single-use | |
Test Tube |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Falcon™ Round-Bottom Polystyrene Test Tubes