missing translation for 'onlineSavingsMsg'
Learn More

Pohli™ Fall Protection Packaging (SUPER FSP)

Approved as combination packaging for self-supporting transport of hazardous goods according to 6PH1/X2.2/250.

Brand:  Pohli™ 03048176

Product Code. 15841418

  • 361.00 € / Each
Estimated Shipment: 06-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Composite packaging SUPER FSP, (UN)6PH1/X2.2/250/JJ/D/ BAM8435/pohli
  • 25L carboy (EG 5 kg flint glass)
  • 48 mm black belts (Polyester) with 2 black handles (HDPE)
  • Mounted DIN 70 black tamper evident screw closure (Lupolen 4261 A)
  • 2mm EPE-liner with 0,2 mm PTFE-coating
  • Green tamper-evident ring (PP)
Specifications

Specifications

(UN)6PH1/X2.2/250/JJ/D/ BAM8435/pohli
435 x 435 x 685 mm (448 x 448 x 688 mm incl. separate card box)
SUPER FSP
Flint Glass, Foamed Polystyrene
Transportation of dangerous goods
Mounted DIN 70 black tamper evident screw closure (Lupolen 4261 A), 2 mm EPE-liner with 0,2 mm PTFE-coating
25 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pohli™ Fall Protection Packaging (SUPER FSP) > L x W x H: 448 x 448 x 688mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Due to glass disease it may happen that the carboys have inside a crust on the surface. Humidity during storage causes salts to hydrate and leach out of the glass. Upon a reduction of relative humidity, these hydrated salts can then form a crust on the surface. These salts are sodium carbonate, calcium carbonate and silicon dioxide. Therefore we recommend to wash the carboys before filling.