missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Water, nuclease-free

Optimize molecular biology applications with this deionized and 0.22μm membrane-filtered nuclease-free water.

31.17 € - 52.25 €

Specifications

pH 7.6
Grade Molecular Biology
Treatment(s) Not DEPC-Treated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11893933
View Documents
Thermo Scientific™
R0581
4 x 1.25 mL
31.17 €
Pack of 4
In Stock
to see stock.
Edge
Add to basket
 
10870051
View Documents
Thermo Scientific™
R0582
30 mL
52.25 €
30mL
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Nuclease-free water is deionized and 0.22 μm membrane-filtered nuclease-free water. It is ideal for all .

Highlights

  • Free of endo-, exodeoxyribonucleases, ribonucleases, and phosphatases
Specifications

Specifications

7.6
Molecular Biology
Not DEPC-Treated
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.