missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ AX-500 Series Compound Research Microscope

Choice of three optical objectives and three optical heads. Fisherbrand AX-500 Series Compound Research Microscope with cool light LED illumination are designed or university and research laboratory use.

Brand:  Fisherbrand™ 1100104400181

 View more versions of this product

Product Code. 15378834


Description

Description

  • Choice of optical objectives, with wide range of magnification: 4x, 10x, 40x, 100x: Achromat, Super high contrast and Plan achromat
  • Choice of optical heads: Monocular, binocular, trinocular (permits attachment of a digital camera)
  • Ergonomically designed, professional frame with low position coaxial focusing allows one handed operation, making extended use comfortable
  • Adjustable 3W LED illumination with intensity control
  • Built-in low drive mechanical stage with extended post for X and Y control (135mm x 140mm)
  • 18mm eyepieces with rubber eyeshields
  • 30° inclined ergonomic Siedentopf viewing head
Specifications

Specifications

Abbe/Iris
Trinocular
4X, 10X, 40X, 100X
DIN Plan Achromatic
IEC power cord, built in cord hanger, dustcover, allen key
Standard DIN
13.3 x 7.0 x 15.3 in. (34 x 18 x 39 cm)
6.5 kg
18 mm
LED
AX-503PL
Microscope
Inward-facing quadruple revolving
2 year for electrical parts (except bulbs, cords and fuses), five year for optical and mechanical parts
14.3 lb.
IEC 110/220 V (UL/CSA)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ AX-500 Series Compound Research Microscope > Trinocular head; Abbe/Iris condenser; Objectives: Plan Achromat DIN 4X, 10X, 40X, 100X

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.