missing translation for 'onlineSavingsMsg'
Learn More
Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters Product Code.: 15397928
Add to basket

Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters

Product Code. 15397928
2, 200/240 V, 50/60 Hz (CN, EU, UK)
Click to view available options
No. of Places:
1
2
4
Electrical Requirements:
200/240 V, 50/60 Hz (CN, EU, UK)

Product Code. 15397928

Edge
Add to basket
Add to basket

Requires

Unit must be ordered with block(s)

Interchangeable modular blocks, configurable for a range of applications, increase lab versatility.

Fisherbrand™ Isotemp Digital Dry Baths/Block Heaters offer a range of configurations with interchangeable modular blocks for a variety of application needs.

Fisherbrand™ Isotemp Digital Dry Baths/Block Heaters offer a range of configurations with interchangeable modular blocks for a variety of application needs. Designed for basic to advanced applications with a maximum temperature of 130°C, a timer and available in 1, 2 and 4 block sizes.

  • Digital controls and display of time and temperature
  • Advanced internal temperature sensing probe for outstanding temperature accuracy and control
  • Precise temperature control with PID circuit
  • Temperature calibration enables to offset the temperature to desired value
  • Timer allows the user to accurately monitor the heating time
  • Wide range of interchangeable aluminum alloy heat blocks provide versatility and allow for easy cleaning and disinfecting
  • Powder-coated steel body construction ensures durability
  • Built-in over-temperature protection device warrants sample and user safety
  • Temperature deviation adjustment

Applications:

Sample preparation, enzyme preservation, enzyme-substrate reactions, DNA amplification, electrophoresis gel degeneration and serum coagulation

Warranty

2-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, cULus, RoHS
Controller Type PID
Material Powder-coated Steel
No. of Places 2
Plug Type CN, EU, UK
Relative Humidity <80%
Temperature Range (Metric) Ambient +5° to 130°C
Type Dry Bath/Block
Fuse 250V 5A
Heat Up Time ≤ 20 min. 30° to 130°C
Includes Dry bath, CN, EU and UK plugs, block lifter
Temperature Accuracy ≤ ± 0.5°C
Temperature Uniformity ≤ ± 1°C
Timer 0 to 99:59 min or continuous
Warranty 2 Year, Replacement
Dimensions (L x W x H) 12.5 x 7.9 x 3.9 in. (318 x 200 x 100 mm)
Length (English) 12.5 in.
Length (Metric) 318 mm
Width (English) 7.9 in.
Height (English) 3.9 in.
Height (Metric) 100 mm
Weight (English) 7.28 lb.
Weight (Metric) 3.3 kg
Electrical Requirements 200/240 V, 50/60 Hz (CN, EU, UK)
Voltage 200/240 V
Frequency 50/60 Hz
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.