missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Absorption Macro Rectangular Cells with PTFE Lid
Includes: PTFE lid
Description
Fit standard spectrophotometer cell holders
- All cells are 12.5mm wide
- 1, 5 or 10mm path length rectangular cells
- Available with a variety of window materials for wide range of assays
- Available in four materials e.g. Quartz, Optical Quartz, Special Optical Glass and Optical Glass
- Quartz windows for transmission from far ultraviolet through the infrared
- Glass windows for routine work in ultraviolet region and visible region
Specifications
Macro Absorption Cell | |
Optical Glass | |
10mm | |
10 mm | |
45 mm | |
12.5 mm | |
Rectangle |
3.5 mL | |
PTFE | |
4/Pk., 8 Pk./Cs. | |
360 to 2500 nm | |
Standard Spectrophotometer Cell Holder | |
Routine work in ultraviolet region and visible region |
Product Suggestions
Customers who viewed this item also viewed
Fisherbrand Focus on glassware and plasticware
File size: 66.4 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Absorption Macro Rectangular Cells with PTFE Lid > w/PTFE lid; Macro; Optical glass; Two clear windows; 360-2500nm; 3000μL; 4/Pk.; 8 Pk./Cs.
Spot an opportunity for improvement?Share a Content Correction