missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BLAUBRAND™ One-mark Glass Bulb Pipets, Class A, USP, Certified
High contrast blue enamel represents optimum combination of resistance and readability
Brand: BRAND™ 30602
89.40 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Made of AR-GLAS™
- Optimized flow geometry in tip ensures reliable liquid dispensing from pipet
- Fire-polished edges for more durability
- Available in all ISO sizes from 0.5 to 100mL
- Supplied with batch number and an accompanying USP batch certificate in original packaging

Specifications
USP | |
AR-GLAS | |
Bulb Pipet |
A | |
1 mL |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BRAND™ BLAUBRAND™ One-mark Glass Bulb Pipets, Class A, USP, Certified > 1mL
Spot an opportunity for improvement?Share a Content Correction