missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Disposable Cuvettes

Choice of polystyrene or methacrylate

Brand:  Fisherbrand™ 14955126

 View more versions of this product

Product Code. 11944385

  • 322.00 € / Pack of 500
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Fisherbrand™ Disposable Cuvettes are available in choice of polystyrene or methacrylate. Select polystyrene cuvets for assays in visible spectral range (340 to 750nm); for accuracy throughout UV/Vis range (285 to 750nm), choose cuvets made from a superior grade of methacrylate.

 

  • Disposable 10mm pathlength cuvets designed for visible and UV/Vis assays
  • Clear four-sided cuvets are ideal for fluorimetry and nephelometry, as well as spectrophotometry
  • Scale down assay volumes and save on reagents by using semimicro cuvets with capacity to 1.5mL
  • Molded-in arrow indicates direction of transmission on standard and semimicro cuvets—helping assure uniformity
  • Each case contains cuvets with same mold impression number—assuring precision between samples—standard deviation is <±0.005 extinction units between cuvets

 

Order Info

Packaged 100 cuvets per stackable tray; five trays to a case. Outside dimensions: 0.47L x 0.47W x 1.77 in. H (12 x 12 x 45mm).

Specifications

Specifications

Standard Cuvette
4.5 mL
12mm
10 mm
45 mm
12 mm
Single-use
PMMA
500 Case
285 to 750 nm
Spectrophotometers, Fluorometers and Nephelometers
For assays in visible spectral range, accuracy throughout UV/Visible range
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Disposable Cuvettes > Standard; Acrylic

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.