missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Disposable Pipette Basins
Disposable basins offer convenience, fast cleanup.
34.00 € - 502.32 €
Product Code | Brand | Capacity (Metric) | Sterility | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Sterility | Packaging | Price | Quantity & Availability | ||||
17815334
|
Fisherbrand™
13-681-509 |
25 mL | Sterile | Bag |
34.00 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
||||
11988485
|
Fisherbrand™
13681501 |
50 mL | Sterile | Individually Wrapped |
410.00 €
Pack of 80 |
In Stock
Log in to see stock. ![]() |
||||
12369175
|
Fisherbrand™
13681500 |
50 mL | Non-sterile | Bulk |
218.00 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
||||
11978485
|
Fisherbrand™
13681502 |
50 mL | Sterile | Bulk |
502.32 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
||||
12399175
|
Fisherbrand™
13681504 |
100 mL | Sterile | Bulk |
381.00 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
||||
Description
Fisherbrand™ Disposable Pipette Basins are packaged in easy-to-open bags to reduce contamination risks. They offer convenience and fast cleanup.
- Easy-pour corners for quick disposal of excess sample solution
25mL Well Basins
- Dimensions: 5.7L × 2.4W × 0.8 in.H (14.5 × 6.3 × 2.1cm)
- Angled sides and V-shaped bottom permit easy withdrawal of sample solution
- Basin is constructed of clear PVC; Sterile (unless opened)
- Dimensions: 5.75L × 2.75W × 1 in.H (14.0 × 7 × 2.5cm)
- Versatile design allows ample clearance for all types of 4-, 8-, and 12-channel pipetters
- Ideal for extended multichannel pipetting into microtiter plates, tubes, and deep-well blocks
- Dimensions: 5.75L × 3.5W × 1.25 in.H (14.6 × 9 × 3.2cm)
Specifications
Pipetting Reservoir | |
Single-use |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Disposable Pipette Basins