Learn More
Fisherbrand™ Disposable Pipette Basins
Description
Fisherbrand™ Disposable Pipette Basins are packaged in easy-to-open bags to reduce contamination risks. They offer convenience and fast cleanup.
- Easy-pour corners for quick disposal of excess sample solution
25mL Well Basins
- Dimensions: 5.7L × 2.4W × 0.8 in.H (14.5 × 6.3 × 2.1cm)
- Angled sides and V-shaped bottom permit easy withdrawal of sample solution
- Basin is constructed of clear PVC; Sterile (unless opened)
- Dimensions: 5.75L × 2.75W × 1 in.H (14.0 × 7 × 2.5cm)
- Versatile design allows ample clearance for all types of 4-, 8-, and 12-channel pipetters
- Ideal for extended multichannel pipetting into microtiter plates, tubes, and deep-well blocks
- Dimensions: 5.75L × 3.5W × 1.25 in.H (14.6 × 9 × 3.2cm)
Specifications
Clear | |
5.75 x 2.75 x 1 in. (14.6 x 7 x 2.54cm) | |
Sterile | |
1 in. | |
5.75 in. | |
Individually Wrapped | |
7 cm |
50 mL | |
Polystyrene | |
Pipetting Reservoir | |
2.54 cm | |
14.6 cm | |
2.75 in. | |
Single-use |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.