Learn More
Fisherbrand™ Elite™ Adjustable-Volume Pipettes
GREENER_CHOICE
Extremely low plunger forces work longer without fatigue and reduce the risk of RSI.
274.00 € - 315.18 €
Specifications
Autoclavable Autoclavable
Product Line Elite
Finger Style Ergonomic
Secure Tip Attachment Soft touch tip ejection
Warranty 3 Years
View More Specs
Products 10
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
11815762
GREENER_CHOICE
View accessory products View Documents
Fisherbrand™
4680000
0.2 to 2 μL
315.18 €
Each
Estimated Shipment: 22-04-2025
to see stock.
Add to basket
11825762
GREENER_CHOICE
View accessory products View Documents
Fisherbrand™
4680010
0.5 to 5 μL
315.00 €
Each
Estimated Shipment: 07-04-2025
to see stock.
Add to basket
11835762
GREENER_CHOICE
View accessory products View Documents
Fisherbrand™
4680020
1 to 10 μL
299.00 €
Each
Estimated Shipment: 22-04-2025
to see stock.
Add to basket
11845762
GREENER_CHOICE
View accessory products View Documents
Fisherbrand™
4680030
2 to 20 μL
305.00 €
Each
In Stock
to see stock.
Edge
Add to basket
11855762
GREENER_CHOICE
View accessory products View Documents
Fisherbrand™
4680040
5 to 50 μL
274.00 €
Each
In Stock
to see stock.
Edge
Add to basket
11865762
GREENER_CHOICE
View accessory products View Documents
Fisherbrand™
4680050
10 to 100 μL
305.00 €
Each
In Stock
to see stock.
Edge
Add to basket
11875762
GREENER_CHOICE
View accessory products View Documents
Fisherbrand™
4680060
20 to 200 μL
305.00 €
Each
In Stock
to see stock.
Edge
Add to basket
11805772
GREENER_CHOICE
View accessory products View Documents
Fisherbrand™
4680090
1 to 10 mL
305.00 €
Each
In Stock
to see stock.
Edge
Add to basket
11895762
GREENER_CHOICE
View accessory products View Documents
Fisherbrand™
4680080
0.5 to 5 mL
305.00 €
Each
In Stock
to see stock.
Edge
Add to basket
11885762
GREENER_CHOICE
View accessory products View Documents
Fisherbrand™
4680070
100 to 1000 μL
315.00 €
Each
In Stock
to see stock.
Edge
Add to basket
Description
Description
Extremely low plunger forces work longer without fatigue and reduce the risk of RSI.
  • Fully autoclavable — entire pipette can be steam autoclaved
  • Soft-touch tip ejection — a gearing mechanism reducing tip ejection force up to 50%
  • Ergonomic comfortable lightweight handle — comfortable grip and effortless tip attachment
  • Optimal fit with SureOne™ pipette tips — best pipetting performance
  • Super blow-out piston — creates 150% bigger air push than conventional piston for delivery of the micro size drops
Warranty
Three years.
Specifications
Specifications
Autoclavable
Ergonomic
3 Years
Elite
Soft touch tip ejection
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Elite™ Adjustable-Volume Pipettes