missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Hand Tally Counter

For counting blood cells, drops of liquid, inventory items, etc.

Brand:  Fisherbrand™ 90023-67

8.00 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.


Product Code. 11704599

  • 18.85 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

For counting blood cells, drops of liquid, inventory items, etc.

Easily resets to zero.
  • Tallies up to 9999; compact
  • Chrome-finished metal construction for reliability, long life, and hard usage
Specifications

Specifications

Hand Tally Counter
9999
Each
85 g
Digital
Chrome Finished Metal
3 oz.
For counting Blood cells, drops of liquid, inventory items, etc.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Hand Tally Counter > Tallies to 9999

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.