missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Handheld Tape Dispenser

Accommodates various tape widths, on 1 in. cores, for easy dispensing.
Brand: Fisherbrand™ 15942
This item is not returnable.
View return policy
Description
- For 0.50, 0.75, or 1 in. wide (1.3, 1.9, or 2.5cm) tapes on 1 in. (2.5cm) core
- Plastic construction
Order Info
Order tape separately.
Specifications
Tape Dispenser | |
Plastic | |
For 0.5, 0.75, or 1 in. W tapes on 1 in. core | |
Handheld |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Handheld Tape Dispenser > Handheld dispenser; For 14 yd. (13m) rolls
Spot an opportunity for improvement?Share a Content Correction