missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Sterile PETG Media Bottles
GREENER_CHOICE

Reduce the risk of cross contamination with these sterile, disposable media bottles

80.45 € - 125.00 €

Specifications

Color Clear
For Use With (Application) For media preparation and storage
Material PETG
Sterility Sterile
Certifications/Compliance SAL 10-6, USP Class VI
View More Specs

Products 3
Product Code Brand Capacity (Metric) Closure Type Price Quantity & Availability  
Product Code Brand Capacity (Metric) Closure Type Price Quantity & Availability  
12074118
GREENER_CHOICE
View Documents
Fisherbrand™
302019-0125
125 mL Tamper-evident Screw Cap
107.00 €
Pack of 24
In Stock
to see stock.
Edge
Add to basket
 
12084118
GREENER_CHOICE
View Documents
Fisherbrand™
302019-0250
250 mL Tamper-evident Screw Cap
125.00 €
Pack of 24
In Stock
to see stock.
Edge
Add to basket
 
12064118
GREENER_CHOICE
View Documents
Fisherbrand™
302019-0500
500 mL Tamper-evident Screw Cap
80.45 €
Pack of 12
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Reduce the risk of cross contamination with these sterile, disposable media bottles

The Fisherbrand PETG Media Bottle is ideal for media preparation and storage.

 

  • An inexpensive and shatterproof alternative to glass media bottles
  • Nonpyrogenic and noncytotoxic to protect cells
  • Molded-in graduations won't rub off
  • Made from clear, lightweight, polyethylene terephthalate copolyester (PETG) providing impact resistance and excellent gas-barrier properties
  • Closure is is high density polyethylene (HDPE)
  • Bottles packed in a shrink-wrapped tray to stay clean
  • Heat-shrink band around cap and bottle neck provides visual assurance of sterility for individual bottles until band is removed
  • Part numbers ending in "A" are designed without a temper-evident ring

 

Compliance

Certified sterile at SAL 10-6, USP Class VI to ensure biocompatibility.

Specifications

Specifications

Clear
PETG
SAL 10-6, USP Class VI
HDPE
For media preparation and storage
Sterile
Media Bottle
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Sterile PETG Media Bottles

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.