Learn More
Description
Traceable™ Humidity-On-A-Card™ Humidity Monitor ideal for use with bottles, chemicals, plastic bags, electronics, desiccators and storage containers.
- Cards record both the current humidity and the maximum humidity attained using color-change reactions
- Current humidity scale ranges from 0 to 100% RH in 10% increments
- Color changes from blue to bright pink as the humidity levels rise and fall
- Accuracy is ±5% RH at 75°F (24°C)
- Maximum humidity scale uses a nonreversible color reaction to record the highest humidity level to which the card has been exposed (over a 24-hour period)
- Range is 50 to 90% RH in 10% increments
- Accuracy: ±5% RH at 75°F (24°C)
- Each card is individually sealed in a clear moisture-barrier bag with a Humidity Sponge™ desiccant to keep the card dry and to provide for an extended shelf life
Certifications
Includes individually serial-numbered ISO 17025 certificate to indicate traceability to standards provided by NIST.
Specifications
Humidity Monitor | |
±5% RH at 75°F (24°C) | |
0.1 | |
Monitor humidity levels in hard-to-reach or out-of-the-way areas over extended time periods, record both the current humidity and the maximum humidity attained using color-change reactions | |
NIST |
0 to 100% | |
24°C | |
75°F | |
Individually serial-numbered ISO 17025 certificate |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.