missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ Fittings for HPLC Systems

Connect your HPLC capillaries safely and tightly to your SCAT™ system. SCAT™ PFA fittings are chemically resistant and fit Safety Caps and Safety Waste Caps. Available for different capillary sizes.

Brand:  SCAT™ 107047

25.89 EUR valid until 2025-04-30
Use promo code "24211" to get your promotional price.


 View more versions of this product

Product Code. 10282320

  • 32.80 € / Each
Estimated Shipment: 12-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

SCAT fittings are available in the following versions:

  • PFA fitting with integrated ferrule in various colors and diameters
  • Tube connectors in various diameters
Specifications

Specifications

Fitting
Solvent Handling
4 mm Diameter Tube
White
Fitting with Integrated Ferrule (4 mm OD, UNF 5/16 to 24 in.)
PTFE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SCAT™ Fittings for HPLC Systems > Fittings; Color: White; For Use With: 4 mm Diameter Tube; Includes: Fitting with Integrated Ferrule (4 mm OD, UNF 5/16 to 24 in.); Type: Fitting

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.