missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Round Aluminum Cookware

Manufactured from aluminum. Bochem™ Round Aluminum Cookware is disposable.

Brand:  Bochem™ 11610

 View more versions of this product

Product Code. 10142171

  • 30.80 € / Pack of 100
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • aluminum
  • Disposable
  • For single use
Specifications

Specifications

Flat
Single-use
Round
For single use
64 mm
28 mL
Aluminum
Dish
13 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bochem™ Round Aluminum Cookware > Capacity: 28mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.