Learn More
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments are accurate and reproducible over a range of volumes.
Brand:  Thermo Scientific™ 365G1503
71.74 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17864961
  • 72.10 € / Each
Explore more special offers
Description
Description
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments are accurate and reproducible over a range of volumes.
  • Manual syringes in industry-standard size
  • White scale backing for easy reading
  • Solvent-resistant markings
TRUSTED_SUSTAINABILITY
Specifications
Specifications
23 G
Ea.
50 mm
AI/AS 1610 Autosampler
36550040 (Side Hole), 36566485 (Cone)
Gas Tight GC Autosampler Syringe, Removable Needle
50 μL
Cone
Removable
Yes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments >
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.