missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Fluoro-Max™ Fluorescent Carboxylate-Modified Particles

Lower detection limits of fluorescent assays by limiting background interference from relatively short-lived matrix fluorescence.

Brand:  Thermo Scientific™ 93470520010350

 View more versions of this product

Product Code. 11844213

  • 11300.00 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Lower detection limits of fluorescent assays by limiting background interference from relatively short-lived matrix fluorescence with Thermo Scientific™ Fluoro-Max Fluorescent Carboxylate-Modified Particles. Made by dyeing Thermo Scientific™ OptiLink™ carboxylate-modified particles with europium chelate, particles have been specifically designed for membrane or automated fluorometric-based applications. With an extremely broad Stokes shift, Fluoro-Max europium chelate particles help prevent nonspecific fluorescence interference.

Fluoro-Max Fluorescent Particles are made from carboxylate-modified polystyrene particles containing encapsulated europium chelate dye. They are available in standard 0.1, 0.2, or 0.3μm diameters. When these particles excite at 333nm and emit at 613nm, they do so with a very long lifetime of approximately 0.5 milliseconds. In fact, this is about 10,000 to 100,000 times the lifetime of most fluorophores. The extended lifetime of europium chelate particles allows them to be used in a wide range of time resolved fluorescence applications. In addition, they can be counted on to lower the detection limits of fluorescent assays severalfold as they eliminate background interference from relatively short-lived matrix fluorescence.
  • Internally dyed to ensure maximum color brilliance and color saturation, resulting in greater sensitivity and enhanced readability of the assay. Eliminates problems associated with enzyme-based assays
  • Encapsulated dye prevents leaching in aqueous media, and provides optimum surface immunoreactivity
  • Dyeing process causes little or no inhibition of adsorption or covalent couping of immunoglobulin G, resulting in superior binding of protein to the particles
  • Suitable for a variety of applications such as clinical diagnostics, immuno/histological studies and research

Recommended for:

Clinical diagnostics, molecular biology and lateral flow, turbidimetric, and slide aggulation assays

TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.05% Sodium Azide
1% solids
Fluorescent red dyed polystyrene microspheres in water
Polystyrene or Polystyrene with Coploymer Grafted Surface
Europium Chelate
20 A²/group
100 mL
Carboxylate-Modified
0.2 μm
0.2 μm
Carboxylate-modified
Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Shipment at ambient temperature. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.
White
Fluroescent Carboxylate-Modified Particle
Package Insert Sheet containing Certicate of Analysis
1.06g/cm³
Product stable for a minimum of 36 months
<5% CV
0.2 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number253282 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number93470520010350
Lot Number254762 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number93470520010350
Lot Number256456 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number93470520010350
Lot Number260213 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number93470520010350
Lot Number280489_STD.STK_1199 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number93470520010350
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Fluoro-Max™ Fluorescent Carboxylate-Modified Particles > Dia: 0.2μm; 100mL; Europium Chelate; Carboxylate-Modified

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.