missing translation for 'onlineSavingsMsg'
Learn More
Formaldehyde, Extra Pure, Solution 37-41%, SLR, Fisher Chemical™ Product Code.: 10231622

Formaldehyde, Extra Pure, Solution 37-41%, SLR, Fisher Chemical™

Product Code. 10231622
1 L, HDPE plastic bottle
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
HDPE plastic bottle
Plastic container
This item is not returnable. View return policy

Product Code. 10231622

Brand: Fisher Chemical F/1501/PB15

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CH2O, CAS Number-50-00-0, 2.5L, 97 deg.C, CHEBI:16842, Colorless, 30.02g/mol, WSFSSNUMVMOOMR-UHFFFAOYSA-N, formaldehyde, -15 deg.C, 3274, 30.026, HDPE plastic bottle, 3 to 4.2, Liquid, 712, C-O, > 1.0, 2mbar at 20 deg.C

CH2O, CAS Number-50-00-0, 2.5L, 97 deg.C, CHEBI:16842, Colorless, 30.02g/mol, WSFSSNUMVMOOMR-UHFFFAOYSA-N, formaldehyde, -15 deg.C, 3274, 30.026, HDPE plastic bottle, 3 to 4.2, Liquid, 712, C-O, > 1.0, 2mbar at 20 deg.C

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Packaging HDPE plastic bottle
Quantity 1 L
Vapor Pressure 2mbar at 20°C
Vapor Density > 1.0
Physical Form Liquid
Color Colorless
pH 3 to 4.2
Melting Point -15°C
Boiling Point 97°C
Formula Weight 30.02g/mol
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formaldehyde Solution 37%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H370-Causes damage to organs.
  • H314-Causes severe skin burns and eye damage.
  • H350-May cause cancer.
  • H341-Suspected of causing genetic defects.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Formaldehyde, Extra Pure, Solution 37-41%, SLR, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.