missing translation for 'onlineSavingsMsg'
Learn More
Formaldehyde solution, Honeywell™ Product Code.: 16626163

Formaldehyde solution, Honeywell™

Product Code. 16626163 Shop All Honeywell Research Chemicals Products
500 mL
Click to view available options
Quantity:
500 mL
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 16626163

Brand: Honeywell Research Chemicals 252549H500ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS reagent, 37 wt.% in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-00-0, 67-56-1, 7732-18-5
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

CAS Min % ≥30.00%, ≥10.00%, ≥50.00%
CAS Max % <50.00%, <20.00%, <70.00%
Quantity 500 mL
Physical Form Liquid
Grade ACS general use solvents
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Formaldehyde solution, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.