Learn More
Thermo Scientific Chemicals Formaldehyde, 37% in aq. soln., ACS, 36.5-38.0%, stab. with 10-15% methanol
Description
Formaldehyde is used as a denaturant in formaldehyde-agarose gel electrophoresis of RNA. It is also used in fiber board, plywood, cigarette smoke, fuel burning appliances, kerosene space heaters and in photography. It acts as a precursor in the manufacturing of automobiles and is used to make components for transmissions, electrical systems, engine blocks, door panels, axles and brake shoes. It serves as a disinfectant. It is also an ingredient in glues and is used as a preservative in medical laboratories, as an embalming fluid and as a sterilizer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | −15°C |
Color | Colorless |
Density | 1.083 g/mL |
Boiling Point | 97°C |
Flash Point | 60°C (140°F) |
Assay Percent Range | 36.5-38.0% |
Odor | Pungent |
Linear Formula | HCHO |
Refractive Index | 1.3765 |
Quantity | 1 L |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
X28K015 | Certificate of Analysis | 08/11/2024 | 033314.M6,033314.K7,033314.AP,033314.M1,033314.K2 |
X15K032 | Certificate of Analysis | 30/10/2024 | 033314.M6,033314.K7,033314.AP,033314.M1,033314.K2 |
W23K021 | Certificate of Analysis | 05/10/2024 | 033314.M6,033314.K7,033314.AP,033314.M1,033314.K2 |
U13K026 | Certificate of Analysis | 06/09/2024 | 033314.M6,033314.K7,033314.AP,033314.M1,033314.K2 |
U16K005 | Certificate of Analysis | 28/08/2024 | 033314.M6,033314.K7,033314.AP,033314.M1,033314.K2 |
Safety and Handling




- Formaldehyde, 37% in aqueous solution
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 3
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- Specific target organ toxicity after single exposure Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- H226-Flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : Formaldehyde, Methanol
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.