Learn More
Four elements solution - Ca, Mg, K, Na for ICP/MS 1g/L HNO3 matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand: Fisher Chemical MS 010 050
Specifications
Four Elements | |
50 mL | |
1000 mg/L | |
Ca, Mg, K, Na |
Bottle | |
Liquid | |
ICP/MS Calibration Multi-element Standard Solution | |
2% HNO3 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.