Learn More
Thermo Scientific™ Frames and Caps for Immuno Standard Modules
BIOPHARMAGREENER_CHOICE
Easily insert and remove strip plates with solid modules by utilizing these frames and caps for immuno standard modules.
93.70 € - 183.82 €
Specifications
Material Polyethylene
Plate Compatibility Immuno Standard Modules
View More Specs
Products 3
Product Code Brand Color Sterility Product Type Price Quantity & Availability  
Product Code Brand Color Sterility Product Type Price Quantity & Availability  
10224551
GREENER_CHOICE
View Documents
Thermo Scientific™
430082
Clear Non-sterile Strip Cap
183.82 €
Pack of 180
In Stock
to see stock.
Add to basket
10684022
GREENER_CHOICE
View Documents
Thermo Scientific™
430805
Clear Sterile Strip Cap
152.51 €
Pack of 120
Estimated Shipment: 29-05-2025
to see stock.
Add to basket
10524811
BIOPHARMA
View Documents
Thermo Scientific™
460348
White Non-sterile Frame
93.70 €
Pack of 60
In Stock
to see stock.
Add to basket
Description
Description
Easily insert and remove strip plates with solid modules by utilizing Thermo Scientific™ Frames and Caps for Immuno Standard Modules.
  • Polyethylene, stackable frame is marked for lid orientation
Specifications
Specifications
Polyethylene
Immuno Standard Modules
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Frames and Caps for Immuno Standard Modules