missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Storage Boxes

Store a variety of vials and tubes with these fiberboard storage boxes, for use in ultra-low temperature freezers.

6.61 € - 52.56 €

Specifications

Color White
Shape Square
Type Cryo Storage Box
For Use With (Equipment) Thermo Scientific Ultra-Low Temperature Freezers
View More Specs

Products 7
Product Code Brand Holds Dimensions (L x W x H) Price Quantity & Availability  
Product Code Brand Holds Dimensions (L x W x H) Price Quantity & Availability  
11976924
View Documents
Thermo Scientific™
5956
Cryo Tubes 5 x 5 x 3 in.
52.56 €
Pack of 12
In Stock
to see stock.
Edge
Add to basket
 
10696335
View Documents
Thermo Scientific™
820002
81 Cryo Tubes 5 x 5 x 2 in.
7.84 €
Each
In Stock
to see stock.
Add to basket
 
11814560
View Documents
Thermo Scientific™
820003
81 Cryo Tubes 5 x 5 x 3 in.
6.61 €
Each
In Stock
to see stock.
Add to basket
 
11349341
View Documents
Thermo Scientific™
820109
100 Cryo Tubes 5 x 5 x 2 in.
7.94 €
Each
In Stock
to see stock.
Add to basket
 
13487248
View Documents
Thermo Scientific™
820013
100 (3 and 4 mL Cryo Tubes) 5.3 x 5.3 x 3.1 in. N/A N/A N/A
13467248
View Documents
Thermo Scientific™
820011
81 (1, 1.5, 2 mL Cryo Tubes) 5.3 x 5.3 x 2 in.
36.98 €
Each
In Stock
to see stock.
Add to basket
 
13457238
View Documents
Thermo Scientific™
820010
100 (1, 1.5, 2 mL Cryo Tubes) 5.3 x 5.3 x 2 in.
38.29 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
Description

Description

Store a variety of vials and tubes with Thermo Scientific™ Storage Boxes, for use in ultra-low temperature freezers.

  • Sturdy, water-repellent boxes with covers for sample storage
  • Available in 2 and 3 in. high (5.1 and 7.6 cm) with 5.25 x 5.25 in. (13.3 x 13.3 cm) square bases
  • Box dividers included excluding 5954 and 5956
Specifications

Specifications

White
Square
Cryo Storage Box
Thermo Scientific Ultra-Low Temperature Freezers
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Storage Boxes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.