missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Funnel Holder

Funnel Holder designed to hold one funnel.

Brand:  BRAND™ 148500

73.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10768431

  • 76.90 € / Pack of 5
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Suitable for supporting 1 or 2 funnels with top outer diameter of 50 to 120 mm
  • Easily mounted on supports rods with an outer diameter of 8 to 14 mm with a locking nut
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Funnel Holder
Funnel (1)
Polypropylene
Funnel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ Funnel Holder > Holds: 1 Funnel

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.