Learn More
R&L Enterprises™ Bench Mounting Gas Cylinder Support
Reduces danger of accidents from falling cylinders or escaping gases, up to 286mm in diameter. R&L Enterprises™ Bench Mounting Gas Cylinder Supports come with fitting instructions.
Brand:  R&L Enterprises™ 3603
Product Code. 10545784
  • 104.00 € / Each
Edge
Explore more special offers
Description
Description
  • Incorporates two screw clamps and holes for permament fixing
  • Heavy metal bracket with fire-resistant strap and buckle
  • Secured permanently or temporarily to a bench edge up to 70mm thick
Specifications
Specifications
Gas Cylinder Support Bench Mounting
Cylinders up to 230mm dia.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&L Enterprises™ Bench Mounting Gas Cylinder Support > For Use With: Cylinders up to 286mm dia.