Learn More
Thermo Scientific™ GeneBLAzer™ D2-Gqo5-NFAT-bla CHO-K1 Cells
GeneBLAzer™ D2-Gqo5-NFAT-bla CHO-K1 cells contain the human Dopamine Receptor 2 (D2) stably integrated into the GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cell line.
Brand:  Thermo Scientific™ K1708
Product Code. 10592754
  • 16830.00 € / Each
Explore more special offers
Description
Description
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
GeneBLAzer™ D2-Gqo5-NFAT-bla CHO-K1 cells contain the human Dopamine Receptor 2 (D2) stably integrated into the GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cell line. GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cells (Cat. No. K1536) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element and the Chimeric G protein, Gqo5.
The GeneBLAzer™ D2-Gqo5-NFAT-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of apomorphine. In addition, GeneBLAzer™ D2-Gqo5-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
The D2 receptor is found primarily in the brain. There are two forms of D2 which are created by alternate splicing: a short form and a long form. This assay uses the long form, although little difference in ligand binding or signaling has been seen between the two forms.
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number36804 Certificate TypeCertificate of Analysis Date16/04/2017 Catalog NumberK1708
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ GeneBLAzer™ D2-Gqo5-NFAT-bla CHO-K1 Cells > 2 x 106 Cells