Learn More
Thermo Scientific™ GeneBLAzer™ NTSR1 CHO-K1 DA Assay Kit
The GeneBLAzer™ NTSR1 CHO-K1 DA cells contain the human Neurotensin Receptor 1 (NTSR1) stably integrated into the GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cell line.
Brand: Thermo Scientific™ K1773
Description
The GeneBLAzer™ NTSR1 CHO-K1 DA cells contain the human Neurotensin Receptor 1 (NTSR1) stably integrated into the GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. No. K1078) contain a beta-lactamase reporter gene under control of the NFAT response element.
The GeneBLAzer™ NTSR1 CHO-K1 DA cells have been functionally validated for Z' factor and EC50 concentrations of established ligands. In addition, GeneBLAzer™ NTSR1 CHO-K1 DA cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary for each product. Division Arrested cells are irreversibly division arrested using a low-dose treatment of Mitomycin C, and have no apparent toxicity or change in cellular signal transduction.

Specifications
NM_002531 | |
Division-Arrested Cells | |
Fluorescence | |
NTSR1 | |
GeneBLAzer™ | |
1 Kit | |
BLA (Beta-Lactamase) | |
Division-Arrested Cells Assay Kit | |
GeneBLAzer™ |
Cell-based beta lactamase reporter gene | |
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol. Includes: • GeneBLAzer™ NTSR1 CHO-K1 DA cells • LiveBLAzer™-FRET B/G Loading Kit, 70 μg • Solution D • GeneBLAzer™ NTSR1 CHO-K1 DA Assay Protocol • LiveBLAzer™-FRET B/G Loading Protocol • Certificate of Analysis The GeneBLAzer™ NTSR1 CHO-K1 DA cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
|
GPCRs | |
Pool of Stable Cells | |
GPCR (G-Protein Coupled Receptors) | |
End Point | |
NTSR1 | |
U2OS | |
GPCR |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
40839 | Certificate of Analysis | 15/04/2017 | K1773 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.