Learn More
Thermo Scientific™ GeneBLAzer™ T-Rex™ GPR119-CRE-bla CHO-K1 Cells
The GeneBLAzer™ T-Rex™ GPR119-CRE-bla CHO-K1 cells contain the human G-Protein Receptor 119 (GPR119), stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line.
Brand:  Thermo Scientific™ K1486
Product Code. 10512394
  • 16830.00 € / Each
Explore more special offers
Description
Description
The GeneBLAzer™ T-Rex™ GPR119-CRE-bla CHO-K1 cells contain the human G-Protein Receptor 119 (GPR119), (Accession No. NP_848566) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat. No. K1129) contain a beta-lactamase (bla) reporter gene under control of the cAMP response element.
The T-Rex™ GPR119-CRE-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of AR231453(1). In addition, T-Rex™ GPR119-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section.
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1855214 Certificate TypeCertificate of Analysis Date19/07/2017 Catalog NumberK1486
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ GeneBLAzer™ T-Rex™ GPR119-CRE-bla CHO-K1 Cells > 2 x 106 Cells