missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Genomic DNA Purification Kit

Thermo Scientific GeneJET Genomic DNA Purification Kit is designed for rapid and efficient purification of high quality genomic DNA from various mammalian cell culture and tissue samples, whole blood, bacteria and yeast.

Brand:  Thermo Scientific™ K0722

598.50 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.



 View more versions of this product

Product Code. 10410450

  • 798.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific GeneJET Genomic DNA Purification Kit is designed for rapid and efficient purification of high quality genomic DNA from various mammalian cell culture and tissue samples, whole blood, bacteria and yeast.

The kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation.

The standard procedure takes less than 20 minutes following cell lysis and yields purified DNA greater than 30 kb in size. Isolated DNA can be used directly in PCR, Southern blotting and enzymatic reactions.

Highlights
Universal—can be used for both cell and tissue samples from a wide range of sources
Efficient—high yields of high molecular weight genomic DNA
Fast—20 minute procedure following cell lysis
Convenient—spin columns are capped and assembled with collection tubes

Applications
Isolated genomic DNA is ideal for use in common molecular biology procedures, including:
• PCR and qPCR
• FastDigest and conventional restriction digestion
• Southern blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Spin Column
Silica Spin Column
Bacteria, Cells, Rodent Tail, Tissue, Whole Blood, Yeast, Buccal Samples
gDNA
Not High-throughput Compatible (Manual)
Room Temperature
≤30 μg
• 5 x 1.2 mL Proteinase K Solution; store at room temperature until opened, –20°C after opening
• 5 x 1 mL RNase A Solution; store at room temperature until opened, –20°C after opening
• 55 mL Digestion Solution; room temperature
• 2 x 60 mL Lysis Solution; room temperature
• 40 mL Wash Buffer I (concentrated); room temperature
• 40 mL Wash Buffer II (concentrated); room temperature
• 150 mL Elution Buffer; room temperature
• 250 GeneJET Purification Columns with Collection Tubes; room temperature
• 250 Collection Tubes; room temperature

Mini
200 μL
PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization
250 Preps
20 min. (After sample homogenization)
Bacteria: ≤109 cells
Cells: ≤106
Rodent Tail: 0.6 cm (rat), 0.5 cm (mouse)
Tissue: ≤20 mg
Whole Blood: ≤200 μL
Yeast: ≤108 cells
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • GeneJET Genomic DNA Purification Kit

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Proteinase, tritirachium album serine, glycerine, Sodium lauryl sulfate, Guanidine hydrochloride

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.