missing translation for 'onlineSavingsMsg'
Learn More

3M™ General Purpose Vinyl Tape 764

This vinyl tape is a cost-effective choice for a variety of hazard, safety marking, and color-coding applications when you’re not dealing with tough conditions or the need for a long-term clean removal. Comes individually wrapped and conveniently packaged.

119.00 € - 123.00 €

Specifications

Adhesive Type Rubber-based Adhesive
Dimensions (L x W) 33 m x 50 mm
Material Vinyl
Packaging Quantity 12 Rolls
Thickness 0.12 mm
View More Specs

Products 6
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
17771689
View Documents
3M™
7100260286
White
119.00 €
12 rolls
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
 
17781689
View Documents
3M™
7100260287
Red
123.00 €
12 rolls
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
 
17761689
View Documents
3M™
7100260285
Yellow
119.00 €
12 rolls
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
 
17721689
View Documents
3M™
7100260280
Green
123.00 €
12 rolls
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
 
17791689
View Documents
3M™
7100260288
Blue
123.00 €
12 rolls
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
 
17721699
View Documents
3M™
7100260600
Black
119.00 €
12 rolls
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
 
Description

Description

  • Ideal for marking applications in light traffic areas, walls and pipes
  • Use for taping in all non-critical applications
  • Maintains vivid color even when exposed to abrasion and solvents
  • Protects parts, equipment and products from abrasions
  • Bundle and temporarily mark parts, machines, and more
  • Easy-to-apply tape stretches around corners for a reliable, tight fit
  • Fast tack and high holding strength
  • Available in wide variety of colors
  • Can be applied by hand or with floor applicator
Specifications

Specifications

Rubber-based Adhesive
Vinyl
0.12 mm
33 m x 50 mm
12 Rolls
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ General Purpose Vinyl Tape 764

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.