missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oakton™ Glass Beads for Mills
Suitable glass beads for the mini-BeedBeater™ Mill. Cole-Parmer™ Glass Beads for Mills are used to crush and separate small samples from bone to microbial spores.
96.45 € - 104.00 €
Specifications
Type | Beads |
---|---|
Material | Glass |
For Use With (Equipment) | Mini-Bead Beater Glass Mill |
Quantity | 1 lb. |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
13478236
|
Oakton™
36270-52 |
0.1 to 0.15 mm |
97.95 €
1kg |
Estimated Shipment: 30-05-2025
Log in to see stock. |
||||||
15237649
|
Oakton™
36270-50 |
1 mm |
104.00 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
12614267
|
Oakton™
36270-51 |
0.5 mm |
96.45 €
Each |
Estimated Shipment: 30-05-2025
Log in to see stock. |
||||||
Description
Suitable glass beads for the mini-BeedBeater™ Mill. Cole-Parmer™ Glass Beads for Mills are used to crush and separate small samples from bone to microbial spores.
- Available in three different sizes
- Crush samples such as bone and microbial spores
- Handles small samples efficiently—from 100μL to 1μL
- Non-foaming, non-aerosol method preserves enzymes, nucleic acids, and organelles
Specifications
Beads | |
Glass | |
Mini-Bead Beater Glass Mill | |
1 lb. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Oakton™ Glass Beads for Mills