Learn More
Oakton™ Glass Beads for Mills
Suitable glass beads for the mini-BeedBeater™ Mill. Cole-Parmer™ Glass Beads for Mills are used to crush and separate small samples from bone to microbial spores.
Brand: Oakton™ 36270-52
Description
Suitable glass beads for the mini-BeedBeater™ Mill. Cole-Parmer™ Glass Beads for Mills are used to crush and separate small samples from bone to microbial spores.
- Available in three different sizes
- Crush samples such as bone and microbial spores
- Handles small samples efficiently—from 100μL to 1μL
- Non-foaming, non-aerosol method preserves enzymes, nucleic acids, and organelles
Specifications
Beads | |
0.1 to 0.15 mm | |
1 lb. |
Glass | |
Mini-Bead Beater Glass Mill |