Learn More
Gilson™ PIPETMAN™ L Mechanical Pipet
GREENER_CHOICE
The PIPETMAN™ L Mechanical Pipet is a lightweight pipet with lockable volume and low forces.
327.00 € - 360.88 €
Spezifikation
Adjustable Tip Spacing No
Autoclavable Autoclavable
Disposable Reusable
Grip Type Ergonomic
Material Polypropylene
Mehr Spezifikationen ansehen
Products 13
Produktcode Marke Volume (Metric) Includes Preis Menge & Verfügbarkeit  
Produktcode Marke Volume (Metric) Includes Preis Menge & Verfügbarkeit  
12366142
GREENER_CHOICE
missing translation for 'viewDocuments'
Gilson™
FA10001M
0.2 to 2 μL Stainless Steel Tip Ejector
327.00 €
Each
In Stock
to see stock.
Edge
Zum Warenkorb hinzufügen
12396142
GREENER_CHOICE
missing translation for 'viewDocuments'
Gilson™
FA10002P
1 to 10 μL Plastic Tip Ejector
336.00 €
Each
In Stock
to see stock.
Zum Warenkorb hinzufügen
12306152
GREENER_CHOICE
missing translation for 'viewDocuments'
Gilson™
FA10003M
2 to 20 μL Stainless Steel Tip Ejector
327.00 €
Each
In Stock
to see stock.
Edge
Zum Warenkorb hinzufügen
12376142
GREENER_CHOICE
missing translation for 'viewDocuments'
Gilson™
FA10001P
0.2 to 2 μL Plastic Tip Ejector
336.00 €
Each
In Stock
to see stock.
Zum Warenkorb hinzufügen
12386142
GREENER_CHOICE
missing translation for 'viewDocuments'
Gilson™
FA10002M
1 to 10 μL Stainless Steel Tip Ejector
360.88 €
Each
In Stock
to see stock.
Edge
Zum Warenkorb hinzufügen
12326152
GREENER_CHOICE
missing translation for 'viewDocuments'
Gilson™
FA10004M
10 to 100 μL Stainless Steel Tip Ejector
327.00 €
Each
Estimated Shipment: 02-07-2025
to see stock.
Zum Warenkorb hinzufügen
12336152
GREENER_CHOICE
missing translation for 'viewDocuments'
Gilson™
FA10004P
10 to 100 μL Plastic Tip Ejector
336.00 €
Each
In Stock
to see stock.
Zum Warenkorb hinzufügen
12346152
GREENER_CHOICE
missing translation for 'viewDocuments'
Gilson™
FA10005M
20 to 200 μL Stainless Steel Tip Ejector
327.00 €
Each
In Stock
to see stock.
Edge
Zum Warenkorb hinzufügen
12356152
GREENER_CHOICE
missing translation for 'viewDocuments'
Gilson™
FA10005P
20 to 200 μL Plastic Tip Ejector
336.00 €
Each
In Stock
to see stock.
Zum Warenkorb hinzufügen
12366152
GREENER_CHOICE
missing translation for 'viewDocuments'
Gilson™
FA10006M
100 to 1000 μL Stainless Steel Tip Ejector
336.00 €
Each
In Stock
to see stock.
Edge
Zum Warenkorb hinzufügen
12386152
GREENER_CHOICE
missing translation for 'viewDocuments'
Gilson™
FA10007
500 to 5000 μL No Tip Ejector
327.00 €
Each
In Stock
to see stock.
Edge
Zum Warenkorb hinzufügen
12376152
GREENER_CHOICE
missing translation for 'viewDocuments'
Gilson™
FA10006P
100 to 1000 μL Plastic Tip Ejector
336.00 €
Each
In Stock
to see stock.
Zum Warenkorb hinzufügen
12396152
GREENER_CHOICE
missing translation for 'viewDocuments'
Gilson™
FA10008
1 to 10 mL No Tip Ejector
349.44 €
Each
In Stock
to see stock.
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Mechanical pipette combines the legendary robustness of the PIPETMAN™ Classic with a newly designed body and state-of-the-art internal mechanism to offer the superior comfort and accuracy. Available in 8 single channel models covering the volume range of 0.2μL to 10mL.
  • Redesigned handle fits naturally in user's hand, no matter the size.
  • Volume locking system guarantees no accidental change in volume while pipetting.
  • Adjustable tip ejector provides comfort for right- and left-handed users.
  • 2D identification code allows easy tracking of product information.
  • New piston assembly designed to drastically improve pipetting and purge forces.
  • Redesigned tip ejector shape and internal mechanism offers reduced tip ejection forces by 50%.
Bestellinformation
Trade in program available, please contact your Fisher Scientific Sales representative for more details.
Spezifikation
Spezifikation
No
Reusable
Polypropylene
Pipetman L
Non-sterile
Ergonomic
1 Year
11 cm
Autoclavable
Ergonomic
1
No
Mechanical Pipette
Transporting a measured volume of liquid
4.33 in.
Certified Quality
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Gilson™ PIPETMAN™ L Mechanical Pipet
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt