Learn More
Hirschmann™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation are manufactured from durable DURAN™ glass tubing.
Brand:  Hirschmann™ 2820181
 View more versions of this product
Product Code. 11772142
  • 70.80 € / Pack of 2
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • With NS ground and polyethylene stopper
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Specifications
DIN EN ISO 1042
100 mL
170 mm
14/23 NS Standard Ground
Yes
Polyethylene
Borosilicate Glass, Duran Glass
±0.1 mL
±0.1 mL
A
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hirschmann™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation > Capacity: 100mL; Socket Joint: 14/23 NS Standard Ground