Learn More
Hirschmann™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation are manufactured from durable DURAN™ glass tubing.
37.65 € - 316.00 €
Specifications
Certifications/Compliance | DIN EN ISO 1042 |
---|---|
Closure Material | Polyethylene |
Graduated | Yes |
Class | A |
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Tolerance | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Tolerance | Price | Quantity & Availability | ||||
10218211
|
Hirschmann™
2820198 |
10 L | 45/40 NS Standard Ground | ±2 mL |
316.00 €
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||
10689451
|
Hirschmann™
2820192 |
1000 mL | 24/29 NS Standard Ground | ±0.6 mL |
61.95 €
Each |
In Stock
Log in to see stock. |
||||
10522891
|
Hirschmann™
2820193 |
1000 mL | 24/29 NS Standard Ground | ±0.4 mL |
67.45 €
Each |
In Stock
Log in to see stock. |
||||
11780964
|
Hirschmann™
2820180 |
100 mL | 12/21 NS Standard Ground | ±0.1 mL |
63.35 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
11760964
|
Hirschmann™
2820161 |
10 mL | 10/19 NS Standard Ground | ±0.04 mL |
54.90 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
11708071
|
Hirschmann™
2820160 |
10 mL | 7/16 NS Standard Ground | ±0.025 mL |
37.65 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
10561161
|
Hirschmann™
2820194 |
2000 mL | 29/32 NS Standard Ground | ±0.6 mL |
106.00 €
Each |
In Stock
Log in to see stock. |
||||
11782142
|
Hirschmann™
2820185 |
200 mL | 14/23 NS Standard Ground | ±0.15 mL |
77.60 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
11718071
|
Hirschmann™
2820165 |
20 mL | 10/19 NS Standard Ground | ±0.04 mL |
57.00 €
Pack of 2 |
In Stock
Log in to see stock. ![]() |
||||
11792142
|
Hirschmann™
2820186 |
250 mL | 14/23 NS Standard Ground | ±0.15 mL |
55.05 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
11816993
|
Hirschmann™
2820171 |
25 mL | 12/21 NS Standard Ground | ±0.06 mL |
43.15 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
11728071
|
Hirschmann™
2820170 |
25 mL | 10/19 NS Standard Ground | ±0.04 mL |
39.20 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
10244712
|
Hirschmann™
2820166 |
20 mL | 12/21 NS Standard Ground | ±0.04 mL |
39.20 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
11702152
|
Hirschmann™
2820190 |
500 mL | 19/26 NS Standard Ground | ±0.25 mL |
95.00 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
10646111
|
Hirschmann™
2820197 |
5 L | 34/35 NS Standard Ground | ±1.2 mL |
168.00 €
Each |
In Stock
Log in to see stock. |
||||
11770964
|
Hirschmann™
2820178 |
50 mL | 14/23 NS Standard Ground | ±0.08 mL |
61.50 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
11750964
|
Hirschmann™
2820153 |
5 mL | 10/19 NS Standard Ground | ±0.04 mL |
52.85 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
11738071
|
Hirschmann™
2820175 |
50 mL | 12/21 NS Standard Ground | ±0.06 mL |
42.75 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
11772142
|
Hirschmann™
2820181 |
100 mL | 14/23 NS Standard Ground | ±0.1 mL |
70.80 €
Pack of 2 |
In Stock
Log in to see stock. ![]() |
||||
11782412
|
Hirschmann™
2820152 |
5 mL | 7/16 NS Standard Taper Ground | 0.025 mL |
37.65 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
Description
- With NS ground and polyethylene stopper
- Acc. to DIN EN ISO 1042
- Corrosion-free and resistant to thermal shock
- Conformity certified, with dated batch identification
Specifications
DIN EN ISO 1042 | |
Polyethylene | |
Yes | |
A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Hirschmann™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation