Learn More
Hirschmann™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation are manufactured from durable DURAN™ glass tubing.
Brand: Hirschmann™ 2820180
This item is not returnable.
View return policy
Description
- With NS ground and polyethylene stopper
- Acc. to DIN EN ISO 1042
- Corrosion-free and resistant to thermal shock
- Conformity certified, with dated batch identification
Specifications
DIN EN ISO 1042 | |
100 mL | |
170 mm | |
12/21 NS Standard Ground | |
Yes |
Polyethylene | |
Borosilicate Glass, Duran Glass | |
±0.1 mL | |
±0.1 mL | |
A |
Product Content Correction
Product Title
Hirschmann™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation > Capacity: 100mL; Socket Joint: 12/21 NS Standard Ground
Spot an opportunity for improvement?Share a Content Correction