Learn More
Marienfeld Superior™ Glass Capillary Tubes for the Determination of Melting Point
Use Marienfeld Superior™ Glass Capillary Tubes for storage of small amounts of liquids and to determine melting points during laboratory experimentation.
Brand:  Marienfeld Superior™ 2930203
 View more versions of this product
Product Code. 10222413
  • 190.74 € / Pack of 1000
Explore more special offers
Description
Description
  • With rectangular cut edges
  • For single use
  • 100 capillaries in a dispenser vial, 10 vials in a box
Specifications
Specifications
Glass
0.42 mm
80 mm
Capillary Tubes (Both Ends Open)
For Determination Of Melting Point
1000
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Marienfeld Superior™ Glass Capillary Tubes for the Determination of Melting Point > Diameter: 0.42mm; Both Ends Open